2-Chloro-5-(trifluoromethyl)phenyl isocyanate ≥97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 97%
CAS No.: 50528-86-4
Molecular Formula: C8H3ClF3NO
Formula Weight: 221.57
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.4890
Boiling Point: 52-53°/3mm
Beilstein Reference: 2112804
UN No.: 2206
MDL No.: MFCD00037029
Formula:
C₈H₃ClF₃NO Punto di ebollizione: 52…53 °C (3 mmHg) Densità: 1,480 Flash Pt: 92 °C (197 °F) |
MDL Number:
MFCD00037029 CAS Number: 50528-86-4 UN: 2206 ADR: 6.1,III |