Stampa…

Diphenylacetylene ≥99%

Fornitore: Thermo Scientific

A13979.06 A13979.14 A13979.22
A13979.06EA 30.4 EUR
A13979.06 A13979.14 A13979.22
Diphenylacetylene ≥99%
Diphenylacetylene

Purity: 99% CAS No.: 501-65-5 Molecular Formula: C14H10 Formula Weight: 178.23 Storage Temperature: 2°C to 8°C Odour: Odourless Boiling Point: 170°/19mm Beilstein Reference: 606478 Merck Reference: 14,9504 MDL No.: MFCD00004786

Formula: C₁₄H₁₀
Peso molecolare: 178,23 g/mol
Punto di ebollizione: 170 °C (19 mmHg)
Punto di fusione: 58…62 °C
Densità: 0,990
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6
Merck Index: 14,9504

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR