2-Methoxythiophenol ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 7217-59-6
Molecular Formula: C7H8OS
Formula Weight: 140.21
Flash Point: 85°(185°F)
Refractive Index: 1.5918
Odour: Unpleasant
Boiling Point: 225-228°
Beilstein Reference: 2042178
MDL No.: MFCD00004834
Formula:
C₇H₈OS Punto di ebollizione: 225…228 °C Densità: 1,152 Flash Pt: 85 °C (185 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004834 CAS Number: 7217-59-6 EINECS: 230-605-7 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...