2-Fluoro-5-nitrobenzoic acid ≥98%
Fornitore: Thermo Scientific
|
Attenzione
|
B22100.06
B22100.14
B22100.06EA
64.8
EUR
B22100.06
B22100.14
2-Fluoro-5-nitrobenzoic acid ≥98%
2-Fluoro-5-nitrobenzoic acid
Purity: 98%
CAS No.: 7304-32-7
Molecular Formula: C7H4FNO4
Formula Weight: 185.11
Melting Point: 142-144°
Beilstein Reference: 1912835
Formula:
C₇H₄FNO₄ Peso molecolare: 185,11 g/mol Punto di fusione: 142…144 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00134238 CAS Number: 7304-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...