Stampa…

2-Bromo-1,3,5-trichlorobenzene ≥97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 1-Bromo-2,4,6-trichlorobenzene

B25000.06 B25000.14
B25000.06EA 71.4 EUR
B25000.06 B25000.14
2-Bromo-1,3,5-trichlorobenzene ≥97%
2-Bromo-1,3,5-trichlorobenzene

Purity: 97% CAS No.: 19393-96-5 Molecular Formula: C6H2BrCl3 Formula Weight: 260.34 MDL No.: MFCD00060653

Formula: C₆H₂BrCl₃
Peso molecolare: 260,34 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00060653
CAS Number: 19393-96-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR