Stampa…
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%
Fornitore: Thermo Scientific
Attenzione
Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid
A10482.03 A10482.06
A10482.03EA 38.9 EUR
A10482.03 A10482.06
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%
3-Hydroxy-4-methoxycinnamic acid
Purity: 98+% CAS No.: 537-73-5 Molecular Formula: C10H10O4 Formula Weight: 194.19 Melting Point: ca 231° dec. Beilstein Reference: 2212760
Formula: C₁₀H₁₀O₄
Peso molecolare: 194,184 g/mol
Punto di fusione: ∼ 231 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0