Methyl pivalate ≥99%
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Methyl trimethylacetate
Purity: 99%
CAS No.: 598-98-1
Molecular Formula: (CH3)3CCO2CH3
Formula Weight: 116.16
Flash Point: 6°(42°F)
Refractive Index: 1.3900
Melting Point:
Formula:
C₆H₁₂O₂ Peso molecolare: 116,16 g/mol Punto di ebollizione: 100…101 °C Punto di fusione: < –70 °C Densità: 0,875 Flash Pt: 6 °C (42 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008843 CAS Number: 598-98-1 EINECS: 209-959-1 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...