Stampa…

Potassium methanesulphonate ≥99%

Fornitore: Thermo Fisher Scientific

Synonyms: Potassiummethanesulphonate, Methanesulphonic acid potassium salt

39505.14 39505.22 39505.36
39505.14EA 64.8 EUR
39505.14 39505.22 39505.36
Potassium methanesulphonate ≥99%
Potassium methanesulphonate

Purity: 99% CAS No.: 2386-56-3 Molecular Formula: CH3KO3S Formula Weight: 134.19 Physical Form: Lump/powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Beilstein Reference: 3914899 MDL No.: MFCD00070544

Formula: CH₃KO₃S
Peso molecolare: 134.2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00070544
CAS Number: 2386-56-3
EINECS: 219-202-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR