Stampa…

Potassio ter-butilato 12% ∼1 mol/L in THF

Fornitore: TCI
Pericolo

Synonyms: Potassio-terz-butossido

P1619-500ML P1619-100ML
TCIAP1619-500MLEA 82 EUR
TCIAP1619-500ML TCIAP1619-100ML
Potassio ter-butilato 12% ∼1 mol/L in THF
Potassio ter-butilato
Formula: C₄H₉KO
Punto di fusione: 256…258 °C
MDL Number: MFCD00012162
CAS Number: 865-47-4
EINECS: 212-740-3
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbcb37d9a905b31ef1f1e99a5cef48ef4f7af662&vid=9e1a173e0bdce6dd3cd4aaae7a899410228ee46b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green clear liquid to slightly cloudy liquid
Concentration (Neutralization titration) 12.0 to 13.2 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR