Stampa…

Diethyl suberate ≥99%

Fornitore: Thermo Fisher Scientific

Synonyms: Diethyl octanedioate, 1,8-Octanedioic acid diethyl ester, Suberic acid diethyl ester

A14735.18 A14735.30 A14735.36
A14735.18EA 103 EUR
A14735.18 A14735.30 A14735.36
Diethyl suberate ≥99%
Diethyl suberate

Purity: 99% CAS No.: 2050-23-9 Molecular Formula: C12H22O4 Formula Weight: 230.30 Flash Point: 110°(230°F) Refractive Index: 1.4320 Boiling Point: 282-283° Beilstein Reference: 1785500 MDL No.: MFCD00009217

Formula: C₁₂H₂₂O₄
Peso molecolare: 230.3 g/mol
Punto di ebollizione: 282…283 °C
Punto di fusione: 5
Densità: 0.980
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009217
CAS Number: 2050-23-9
EINECS: 218-084-4

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR