Acido succinico ≥99.0% ACS
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
1,4-Butanedioic acid
Purity: 99.0% min
CAS No.: 110-15-6
Molecular Formula: HO2CCH2CH2CO2H
Formula Weight: 118.09
Flash Point: 206°(402°F)
Physical Form: Crystalline
Density: 1.56
Melting Point: 186-188°
Boiling Point: 235°
Solubility: Soluble in water (increasing with temperature). Soluble in alcohol, methanol
Beilstein Reference: 1754069
Merck Reference: 14,8869
MDL No.: MFCD00002789
Formula:
HOOCCH₂CH₂COOH Punto di ebollizione: 235 °C Punto di fusione: 186…188 °C Densità: 1,56 Flash Pt: 206 °C (402 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 EINECS: 203-740-4 UN: 3261 ADR: 8,III Merck Index: 14,8869 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...