1,15-Pentadecanedioic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
P0909-1G
P0909-10G
TCIAP0909-1GEA
23
EUR
TCIAP0909-1G
TCIAP0909-10G
1,15-Pentadecanedioic acid ≥97.0% (by GC, titration analysis)
1,15-Pentadecanedioic acid
Formula:
C₁₅H₂₈O₄ Punto di ebollizione: 175°C |
MDL Number:
MFCD00039534 CAS Number: 1460-18-0 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 113.0 to 116.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...