Ethyl laurate ≥98%
Fornitore: Thermo Scientific
Synonyms:
Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester
Purity: 98%
CAS No.: 106-33-2
Molecular Formula: CH3(CH2)10CO2CH2CH3
Formula Weight: 228.38
Flash Point: 138°(280°F)
Refractive Index: 1.4315
Odour: Fruit-like
Melting Point: -2°
Boiling Point: 268-270°
Beilstein Reference: 1769671
Merck Reference: 14,3818
MDL No.: MFCD00015065
Formula:
C₁₄H₂₈O₂ Peso molecolare: 228,38 g/mol Punto di ebollizione: 268…270 °C Punto di fusione: –2 °C Densità: 0,862 Flash Pt: 138 °C (280 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00015065 CAS Number: 106-33-2 EINECS: 203-386-0 Merck Index: 14,3818 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...