N,N-Dietilformamide ≥99%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 99%
CAS No.: 617-84-5
Molecular Formula: HCON(CH2CH3)2
Formula Weight: 101.15
Flash Point: 65°(149°F)
Refractive Index: 1.4340
Odour: Amine-like
Boiling Point: 178°
Beilstein Reference: 1209392
MDL No.: MFCD00003287
Formula:
C₅H₁₁NO Punto di ebollizione: 178 °C Densità: 0,910 Flash Pt: 65 °C (149 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003287 CAS Number: 617-84-5 EINECS: 210-533-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...