Valeryl chloride ≥98%
Fornitore: Thermo Fisher Scientific
Synonyms:
Pentanoyl chloride, Valeroyl chloride
Purity: 98%
CAS No.: 638-29-9
Molecular Formula: CH3(CH2)3COCl
Formula Weight: 120.58
Flash Point: 32°(89°F)
Refractive Index: 1.4200
Odour: Acrid
Melting Point: -110°
Boiling Point: 125-127°
Beilstein Reference: 506293
UN No.: 2502
MDL No.: MFCD00000757
Formula:
C₅H₉ClO Punto di ebollizione: 125 °C (1013 hPa) Punto di fusione: –110 °C Densità: 1,016 g/cm³ (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00000757 CAS Number: 638-29-9 EINECS: 211-330-1 UN: 2502 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...