Stampa…

5-Bromovaleryl chloride ≥98%

Fornitore: Thermo Fisher Scientific
Pericolo

A18046.14 A18046.06
A18046.14EA 240 EUR
A18046.14 A18046.06
5-Bromovaleryl chloride ≥98%
5-Bromovaleryl chloride

Purity: 98% CAS No.: 4509-90-4 Molecular Formula: C5H8BrClO Formula Weight: 199.46 Flash Point: 107°(225°F) Refractive Index: 1.4910 Odour: Irritating Boiling Point: 43-45°/0.2mm Beilstein Reference: 1745663 UN No.: 3265 MDL No.: MFCD00013660

Formula: BrH₂C(CH₂)₃COOCl
Punto di ebollizione: 43…45 °C (0,2 mmHg)
Densità: 1,49
Flash Pt: 107 °C (225 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00013660
CAS Number: 4509-90-4
UN: 3265
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR