Stampa…

(R)-(-)-α-Methoxyphenylacetic acid ≥99%

Fornitore: Thermo Scientific
Attenzione

Synonyms: (R)-(-)-alpha-Methoxyphenylacetic Acid

L08719.03 L08719.MD
L08719.03EA 81.9 EUR
L08719.03 L08719.MD
(R)-(-)-α-Methoxyphenylacetic acid ≥99%
(R)-(-)-α-Methoxyphenylacetic acid

Purity: 99% CAS No.: 3966-32-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 63-68° Boiling Point: 164-166°/18mm MDL No.: MFCD00004250

Formula: C₉H₁₀O₃
Peso molecolare: 166,18 g/mol
Punto di ebollizione: 164…166 °C (18 mmHg)
Punto di fusione: 63…68 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR