Stampa…
Rame (II) nitrato 2,5-idrato ≥97%, TECHNICAL
Fornitore: VWR Chemicals
Pericolo
Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate
23121.297EA 94 EUR
23121.297 23121.366
Rame (II) nitrato 2,5-idrato ≥97%, TECHNICAL
Rame (II) nitrato 2,5-idrato
Formula: Cu(NO₃)₂·2.5H₂O
Peso molecolare: 232.59 g/mol
Punto di fusione: 114 °C
Densità: 2.05 g/cm³ (20 °C)
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 1477
ADR: 5.1 II
Visualizza i codici
Assay Min. 97 %