Europium(III) chloride hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Fornitore: Thermo Scientific
Synonyms:
Europium chloride hydrate, Europium trichloride hydrate
Purity: 99.9%
Molecular Formula: EuCl3.xH2O
Formula Weight: 258.32(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Odour: Odourless
Merck Reference: 14,3904
MDL No.: MFCD00010994
Formula:
Cl₃Eu·xH₂O Peso molecolare: 258,32 g/mol (anhy) Punto di fusione: 158 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00217047 EINECS: 233-040-4 Merck Index: 14,3904 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...