Stampa…
1-Octanesulphonic acid sodium salt monohydrate
Fornitore: MP Biomedicals
Synonyms: Sodium 1-octansulphonate monohydrate, Sodium 1-octanesulphonate monohydrate
0215177805 0215177825 0215177880
ICNA0215177805EA 39.1 EUR
ICNA0215177805 ICNA0215177825 ICNA0215177880
1-Octanesulphonic acid sodium salt monohydrate
1-Octanesulphonic acid sodium salt monohydrate
Formula: C₈H₁₇NaO₃S·1H₂O
Peso molecolare: 234,29 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007544
CAS Number: 207596-29-0
EINECS: 226-195-4