Stampa…

1-Amino-2,2,2-trifluoroethane hydrochloride 98%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 2,2,2-Trifluoroethanamine hydrochloride, 2,2,2-Trifluoroethylamine hydrochloride

A14904.09 A14904.18 A14904.22
A14904.09EA 84.1 EUR
A14904.09 A14904.18 A14904.22
1-Amino-2,2,2-trifluoroethane hydrochloride 98%
1-Amino-2,2,2-trifluoroethane hydrochloride

Purity: 98% CAS No.: 373-88-6 Molecular Formula: C2F3H4N.HCl Formula Weight: 135.52 Melting Point: 218° subl. Beilstein Reference: 3652103

Formula: C₂H₅ClF₃N
Peso molecolare: 135,52 g/mol
Punto di fusione: 218 °C (subl.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00012875
CAS Number: 373-88-6
EINECS: 206-771-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR