Stampa…
Acido cloridrico concentrate for dilution to 1L, 0.2 M HCl (0.2N) soluzione volumetrica, Fluka™
Fornitore: Honeywell Chemicals
Pericolo
Synonyms: Hydrochloric acid solution, Muriatic acid
38287-1EA 38287-6X1EA
38287-1EAEA 63.01 EUR
38287-1EA 38287-6X1EA
Acido cloridrico concentrate for dilution to 1L, 0.2 M HCl (0.2N) soluzione volumetrica, Fluka™
Acido cloridrico
Concentrate for 1L standard solution
Formula: HCl
Peso molecolare: 36.46 g/mol
Densità: 1.11 g/cm³
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
Visualizza i codici
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective