(±)-2-Mercaptopropionic acid ≥97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
2-Mercaptopropionic acid
Purity: 97%
CAS No.: 79-42-5
Molecular Formula: C3H6O2S
Formula Weight: 106.14
Flash Point: 107°(225°F)
Refractive Index: 1.4820
Odour: Unpleasant
Melting Point: 10-14°
Boiling Point: 217°
Beilstein Reference: 506218
Merck Reference: 14,9340
UN No.: 2936
MDL No.: MFCD00004862
Formula:
C₃H₆O₂S Peso molecolare: 106.15 g/mol Punto di ebollizione: 217 °C Punto di fusione: 10…14 °C Densità: 1.196 Flash Pt: 107 °C (225 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004862 CAS Number: 79-42-5 EINECS: 201-206-5 UN: 2936 ADR: 6.1 II Merck Index: 12,09476 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...