Stampa…

3-Bromopropionic acid ≥97%

Fornitore: Thermo Scientific
Pericolo

Synonyms: 3-Bromopropanoic acid

A11573.14 A11573.22 A11573.36
A11573.14EA 28.3 EUR
A11573.14 A11573.22 A11573.36
3-Bromopropionic acid ≥97%
3-Bromopropionic acid

Purity: 97% CAS No.: 590-92-1 Molecular Formula: BrCH2CH2CO2H Formula Weight: 152.98 Flash Point: 65°(149°F) Melting Point: 57-63° Boiling Point: 140-142°/45mm Solubility: Soluble in water, alcohol, ether, chloroform, benzene Beilstein Reference: 1071333 Merck Reference: 14,1433 UN No.: 3261 MDL No.: MFCD00002763

Formula: BrH₂CCH₂COOH
Peso molecolare: 152,98 g/mol
Punto di ebollizione: 140…142 °C (45 mmHg)
Punto di fusione: 57…63 °C
Densità: 1,480
Flash Pt: 65 °C (149 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002763
CAS Number: 590-92-1
EINECS: 209-694-1
UN: 3261
ADR: 8,II
Merck Index: 14,1433

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR