Stampa…
Tween® 20 (Polysorbate), EMPROVE® ESSENTIAL JPE, Ph. Eur., NF suitable for use as excipient, SAFC®
Fornitore: MERCK PRODUCTION CHEMICALS
Synonyms: Polyoxyethylene sorbitan monolaurate
8.17072.1000 8.17072.2500
8.17072.1000EA 330 EUR
8.17072.1000 8.17072.2500
Tween® 20 (Polysorbate), EMPROVE® ESSENTIAL JPE, Ph. Eur., NF suitable for use as excipient, SAFC®
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
Peso molecolare: 1227.53 g/mol
Densità: 1.1 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3