Platinum rhodium alloy (94:6 w%), in filo for ISA type B standard grade thermocouple, Ø 0.25 mm (0.01 in)
Fornitore: Thermo Fisher Scientific
Molecular Formula: Pt:Rh; 94:6 wt%
Physical Form: ≈0.52g/50cm
Odour: Odourless
MDL No.: MFCD00198186