4-Hydroxychalcone ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 20426-12-4
Molecular Formula: C15H12O2
Formula Weight: 224.26
Melting Point: 183-185°
Beilstein Reference: 2049155
Formula:
C₁₅H₁₂O₂ Punto di fusione: 183…185 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00016488 CAS Number: 20426-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...