Stampa…
Cobalto ≥99.998% (metals basis), in polvere, Puratronic® -22 mesh
Fornitore: Thermo Scientific
Pericolo
10610.06 10610.14 10610.A1
10610.06EA 106 EUR
10610.06 10610.14 10610.99
Cobalto ≥99.998% (metals basis), in polvere, Puratronic® -22 mesh
Cobalto
Purity: 99.998% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935
Formula: Co
Peso molecolare: 58,93 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,III