Thulium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Fornitore: Thermo Scientific
|
Pericolo
|
confezionato sotto argo
12435.03
12435.06
12435.03EA
131
EUR
12435.03
12435.06
Thulium ≥99.9% (REO, rare earth oxide basis), pieces, sublimed dendritic
Thulium
Purity: 99.9%
CAS No.: 7440-30-4
Physical Form: Packaged under argon
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00011281
Formula:
Tm Peso molecolare: 168,93 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00011281 CAS Number: 7440-30-4 EINECS: 231-140-2 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...