Stampa…
Zolfo, meets analytical specification of Ph.Eur., per uso esterno, SAFC®
Fornitore: MERCK PRODUCTION CHEMICALS
Attenzione
1.07983.9025
1.07983.9025EA 615 EUR
1.07983.9025
Zolfo, meets analytical specification of Ph.Eur., per uso esterno, SAFC®
Zolfo
Formula: S
Peso molecolare: 32,07 g/mol
Punto di ebollizione: 445 °C (1013 hPa)
Punto di fusione: 113…119 °C
Densità: 2,36 g/cm³ (20 °C)
Flash Pt: 168 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059
Visualizza i codici
Meets analytical specifications of Sulfur for external use Ph Eur.
Assay 99.0 - 101.0 %
Identity passes test
Appearance of solution passes test
Odour passes test
Acidity or alkalinity passes test
Chloride (Cl) ≤ 0.0100 %
Sulfate (SO₄) ≤ 0.0100 %
Sulfide (S) passes test
Arsenic,Selenium (as As) ≤ 0.0004 %
Residual solvents (ICH Q3C) excluded by production process
Sulfated ash (600 °C) ≤ 0.2 %
The information provided does not imply the suitability of the product for any particular application. It is customer’s sole responsibility, prior to use, to determine that the product is suitable and permitted for the customer’s intended use and application.