Stampa…

Silicone ≥99.5% (metals basis), crystalline powder -325 mesh

Fornitore: Thermo Scientific
Attenzione

12681.09 12681.22 12681.36
12681.09EA 54.8 EUR
12681.09 12681.22 12681.36
Silicone ≥99.5% (metals basis), crystalline powder -325 mesh
Silicone

Purity: 99.5% CAS No.: 7440-21-3 Appearance: White Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
Peso molecolare: 28,09 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR