Magnesio ≥99.98% (metals basis), trucioli, Puratronic® 1 cm (0.4 in) & down
Fornitore: Thermo Scientific
|
Pericolo
|
10630.06
10630.14
10630.22
10630.36
10630.06EA
30.7
EUR
10630.06
10630.14
10630.22
10630.36
Magnesio ≥99.98% (metals basis), trucioli, Puratronic® 1 cm (0.4 in) & down
Magnesio
Purity: 99.98%
CAS No.: 7439-95-4
Odour: Odourless
UN No.: 1869
MDL No.: MFCD00085308
Formula:
Mg Peso molecolare: 24,3 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...