Stampa…
Beakers, borosilicate glass, with graduation
Fornitore: JONES FR
BN.351 BN.352 BN.353 BN.354 BN.355 BN.356 BN.358 BN.370 BN.373
JONEBN.351EA 0 EUR
JONEBN.351 JONEBN.352 JONEBN.353 JONEBN.354 JONEBN.355 JONEBN.356 JONEBN.358 JONEBN.370 JONEBN.373
Beakers, borosilicate glass, with graduation
Becher
These beakers are made of borosilicate glass in accordance with BS.1922. Each measure is government stamped.
  • Lighter, aids handling especially in the larger sizes
  • With permanent enamel graduations
  • Very good chemical and temperature resistance
The flat bottom allows for easier stirring on hotplates and homogenising. The calibration line spacing and accuracy are the same as traditional conical and bell measures.