Stampa…
Jars, waste disposal
Fornitore: MEDLINE SCIENTIFIC
870004 870004H 870036 870036H
129-0590EA 275 EUR
129-0590 129-0591 129-0592 129-0593
Jars, waste disposal
Contenitori Contenitori, per usi generali
PET, transparent with red PP lid.
  • Small, square jars with a fold-away carrying handle, large circular jars without a carrying handle
  • Food grade quality
  • Available with or without 'Biohazard' label
  • High clarity polyethylene is ideal to autoclave, causing the container to collapse around the waste material and ultimately minimising the space taken for final safe disposal
Dispo jars, often referred to as sweet jars. Ideal for collecting and disposing of waste, such as loops, cotton wool, wipes and other small hazardous waste in every day microbiology or pathology procedures. Also suitable for a wide range of other applications.