Stampa…

MDL-28170

Fornitore: ENZO LIFE SCIENCES

BMLPI1300010 BMLPI1300050
ENZOBMLPI1300010EA 67.1 EUR
ENZOBMLPI1300010 ENZOBMLPI1300050
MDL-28170
Proteine e Peptidi

A potent and selective cell permeable calpain inhibitor. Inhibits oxidative damage-induced apoptosis in PC12 cells and capsaicin-induced apoptosis in dorsal root ganglion neurons. It protects against anoxia in rat optic nerves and reduces neuronal damage in an animal model of global cerebral ischemia(50 mg/kg). In isolated hippocampal pyramidal neurons, it inhibits Ca²⁺-induced suppression of neurite outgrowth (complete inhibition at 20 µM). Calpain I Ki = 8 nM; Cathepsin B Ki = 24 nM in isolated enzyme assays. IC50 = 0,3 µM in intact cell assay.


  • Calpain and cathepsin inhibitor


Soluble in DMSO (200 mM).

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b31ed6b509115c3006261d49c28d0bc2da45591d&vid=e3fe31efb1b5f7f98ee2921449b3300842651266&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR