Stampa…
Ac-YVAD-AMC
Fornitore: ENZO LIFE SCIENCES AG
ALX260024M001 ALX260024M005 ALX260024M001 ALX260024M005
ALEXALX260024M001EA 0 EUR
ALEXALX260024M001 ALEXALX260024M005 ENZOALX260024M001 ENZOALX260024M005
Ac-YVAD-AMC
Proteine e Peptidi
Caspase-1 inhibitor
Fluorgogenic caspase-1 substrate with a Km=14µM. Ex.:340-360nm, Em.:440-460nm.Sequence is based on precursor interleukin-1beta Asp116 cleavage site for caspase-1.