Stampa…
Mouse nerve growth factor 25S (from submaxillary glands)
Fornitore: ENZO LIFE SCIENCES AG
ALX521007C025 ALX521007C100 ALX521007C025 ALX521007C100
ALEXALX521007C025EA 0 EUR
ALEXALX521007C025 ALEXALX521007C100 ENZOALX521007C025 ENZOALX521007C100
Mouse nerve growth factor 25S (from submaxillary glands)
Proteine e Peptidi
Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984).
Informazioni per l'ordine: Quantity: Concentration is estimated spectroscopically from its extinction coefficient and amino acid analysis.