Stampa…
Microcystis aeruginosa Microcystin LW
Fornitore: ENZO LIFE SCIENCES AG
ALX350080C025 ALX350080C025 ALX350080C100
ALEXALX350080C025EA 0 EUR
ALEXALX350080C025 ENZOALX350080C025 ENZOALX350080C100
Microcystis aeruginosa Microcystin LW
Proteine e Peptidi
Microcystins are a group of cyclic heptapeptide hepatotoxins produced by a number of cyanobacterial genera. The most notable of which, and namesake, is the widespread genus Microcystis. Structurally, all microcystins consist of the generalized structure cyclo(-D-Ala1-X2-D-MeAsp3-Y4-Adda5-D-Glu6-Mdha7-). X and Y are variable L-amino acids, D-MeAsp is D-erythro-ß-methylaspartic acid and Mdha is N-methyldehydroalanine. Adda is the cyanobacteria unique C20 ß-amino acid 3-amino-9-methoxy-2,6,8-trimethyl-10-phenyl-deca-4,6-dienoic acid. Substitutions of the variable L-amino acids at positions 2 and 4 give rise to at least 21 known primary microcystin analogs and alterations in the other constituent amino acids result in more than 90 reported mycrocystins to date.
Analog of microcystin-LR (Prod. No. ALX-350-012) with Trp substituted in place of Arg. Hydrophobic and believed to be more cell permeable than other microcystins. May prove useful in biochemical studies in intact cells. Microcystin-LW has a characteristically different absorption spectrum compared to other microcystins, making it a useful reference compound for HPLC analysis. The Trp confers an absorption maximum at 222 nm, whereas most microcystins have a characteristic maximum at 239 nm.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48884c890362272dc9b220d37caafc204f6520a6&vid=11083068c9da50600f0d7f1ec65ee0f9afdce604&prid=web&clid=SAPDAM