Stampa…
Ac-VDVAD-AFC
Fornitore: ENZO LIFE SCIENCES
ALX260112M005 ALX260112M010 ALX260112M005 ALX260112M010
ALEXALX260112M005EA 0 EUR
ALEXALX260112M005 ALEXALX260112M010 ENZOALX260112M005 ENZOALX260112M010
Ac-VDVAD-AFC
Proteine e Peptidi
Caspase-2 substrate
Fluorogenic substrate for caspase-2; similar to Ac-VDVAD-AMC but the AFC fluorophore has a greater Stokes’ shift upon cleavage. VDVAD has been found to be a preferred cleavage site for caspase-2 (Ich-1L). Increases in Ac-VDVAD-AFC cleavage correlated with losses in procaspase-2 (conversion to active caspase-2) in human neuroblastoma lines induced to apoptosis with C2-ceramide and NO.