Stampa…
Ac-DEVD-AFC
Fornitore: ENZO LIFE SCIENCES
ALX260032M001 ALX260032M005 ALX260032M001 ALX260032M005
ALEXALX260032M001EA 0 EUR
ALEXALX260032M001 ALEXALX260032M005 ENZOALX260032M001 ENZOALX260032M005
Ac-DEVD-AFC
Proteine e Peptidi
Caspase-3 substrate
Fluorogenic substrate for caspase-3 (CPP32), with a Km=9.7µM) and related cysteine proteases. Sequence is based on PARP cleavage at Asp216 for caspase-3. Similar to Ac-DEVD-AMC (Prod. No. ALX-260-031) but the AFC fluorophore has a greater Stokes' shift upon cleavage than AMC. Reaction can be monitored quantitatively or visually using a hand-held long-UV lamp and visualizing a blue to green shift in fluorescence upon cleavage. Ex.: 400 nm, Em.: 505 nm.
Visualizza i codici
Specifiche
Protein/peptide name Ac-DEVD-AFC Ac-DEVD-AFC
Environmentally Preferable
Specie
Sorgente luminosa
Purezza >96% (HPLC) >96% (HPLC)