Stampa…
Synthetic melittin
Fornitore: ENZO LIFE SCIENCES
ALX162007M001 ALX162007MC05 ALX162007M001 ALX162007MC05
ALEXALX162007M001EA 0 EUR
ALEXALX162007M001 ALEXALX162007MC05 ENZOALX162007M001 ENZOALX162007MC05
Synthetic melittin
Proteine e Peptidi
PLA2 stimulatory bee venom peptide. Sequence is similar to Phospholipase A2 Activating Peptide (Prod. No. ALX-165-007). Activates PLA2 (5-fold at 10 µg/ml) in BC3H1 cells. For additional technical information see melittin (natural) (Prod. No. ALX-162-006).
  • PLA2 activator