Stampa…
AMPIGENE® qPCR Green Mix Hi-ROX
Fornitore: ENZO LIFE SCIENCES AG
ENZNUC1040200 ENZNUC1041000
ENZOENZNUC1040200EA 124.3 EUR
ENZOENZNUC1040200 ENZOENZNUC1041000
AMPIGENE® qPCR Green Mix Hi-ROX
Reagenti per acidi nucleici Enzimi e kit qPCR/RT-qPCR
Optimised dye-based qPCR.
AMPIGENE® qPCR Green Mixes use the latest developments in polymerase technology and buffer chemistry to deliver high performing dye-based real-time PCR with minimal optimization. AMPIGENE® qPCR Green Mixes use a proprietary intercalating dye that does not inhibit PCR.An optimized buffer system allows efficient amplification and proprietary enhancers prevent the formation of primers/dimers to increase sensitivity and specificity. ROX is a reference dye which is used to normalize fluorescent signal in qPCR. AMPIGENE® qPCR Green Mixes come with either Lo-ROX or Hi-ROX concentration. For Lo-ROX, see Prod. No. ENZ-NUC103. Choose the appropriate ROX concentration for the instrument.
Informazioni per l'ordine: Contains 2x AMPIGENE® qPCR Green Mix Hi-ROX (includes dNTPs and polymerases).