Stampa…
Anti-NAMPT Mouse Monoclonal Antibody [clone: P4D5AT]
Fornitore: ENZO LIFE SCIENCES
ALX804922C050 ALX804922C100
ENZOALX804922C050EA 359 EUR
ENZOALX804922C050 ENZOALX804922C100
Anti-NAMPT Mouse Monoclonal Antibody [clone: P4D5AT]
Anticorpi
Type: Primary
Antigen: NAMPT
Clonality: Monoclonal
Clone: P4D5AT
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human