Stampa…
Anti-HSP90B1 Rabbit Polyclonal Antibody
Fornitore: ENZO LIFE SCIENCES
ADISPA851D ADISPA851F
ENZOADISPA851DEA 229 EUR
ENZOADISPA851D ENZOADISPA851F
Anti-HSP90B1 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: HSP90B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat