Stampa…

Anti-HSP25 + HSP27 Rabbit Polyclonal Antibody

Fornitore: ENZO LIFE SCIENCES
ADISPA525D ADISPA525F
ENZOADISPA525DEA 313 EUR
ENZOADISPA525D ENZOADISPA525F
Anti-HSP25 + HSP27 Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: HSP25 + HSP27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Guinea pig, Dog, Bovine
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR