Stampa…

Anti-Titin Mouse Monoclonal Antibody

Fornitore: ENZO LIFE SCIENCES
ALXBC3010SL001 ALXBC3010SL005 ALXBC3010SL001 ALXBC3010SL005
ALEXALXBC3010SL001EA 0 EUR
ALEXALXBC3010SL001 ALEXALXBC3010SL005 ENZOALXBC3010SL001 ENZOALXBC3010SL005
Anti-Titin Mouse Monoclonal Antibody
Anticorpi
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes human, mouse and rabbit titin.

Type: Primary
Antigen: TITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR