Stampa…
Combined fridge freezers, Premium BioFresh, CBNPes series
Fornitore: Liebherr
CBNPES3756 CBNPES3976
LIEBCBNPES3756EA 0 EUR
LIEBCBNPES3756 LIEBCBNPES3976
Combined fridge freezers, Premium BioFresh, CBNPes series
Refrigeratori
These combined fridge freezers with biofresh and NoFrost offer more interior capacity for food storage, economy and energy efficiency.
  • SwingLine design
  • Stainless steel slimline handles with integrated opening mechanism
  • Versatile installation due to front ventilation
  • Transport handles at back
  • Height-adjustable feet at front, transport castors at back
  • Reversible door hinging
These freezers feature stainless steel door with SmartSteel and aluminium door handle with integrated opening mechanism. The NoFrost system provides rapid, fan-assisted, frost-free freezing for the safe, longterm storage of food. The NoFrost technology creates a much larger storage capacity and keeps the freezer constantly frost-free. The BioFresh safes pull out on smooth-running telescopic rails and can easily be removed once the door is open to an angle of 90°
Accurate and independent temperature control of the fridge and freezer compartments is facilitated by DuoCooling: two separately adjustable cooling circuits. There is no air exchange between the fridge and freezer compartment, so that odours cannot be transferred and the stored food is prevented from drying out.
The high-resolution, high-contrast TFT display is integrated into the door and offers individual setting options. Appliances can be adapted to specific customer requirements using preset programmes, thereby ensuring optimal food storage.
The distinctive GlassLine interior conveys build and finish quality. The shelves are made of safety glass. The sectioned, slide-under glass shelf lends itself to versatile use as it can be inserted at different heights, as required.