Stampa…
Cryoboxes, Cardboard
Fornitore: NEOLAB
2-2883 2-2671 2-2672 2-2673 2-2674 2-2677 2-2880 2-2881 2-2882 2-2884 2-2885 2-2886 2-2887
NEOL2-2883EA 40.3 EUR
NEOL2-2883 NEOL2-2671 NEOL2-2672 NEOL2-2673 NEOL2-2674 NEOL2-2677 NEOL2-2880 NEOL2-2881 NEOL2-2882 NEOL2-2884 NEOL2-2885 NEOL2-2886 NEOL2-2887
Cryoboxes, Cardboard
Scatole Cryobox
These cryoboxes are made of cryo resistant and impregnated cardboard.
  • Pop-up lid without labelling field
For storage of cryovials.