Stampa…
Sensori di conducibilità
Fornitore: Mettler - Toledo
51302255 51302256 51344020 51344022 51344024 51344120 51344122 51344124 51344126 51344128 51344030 51344031 30014160 51340336 51344026
662-2814EA 464 EUR
662-2814 663-0061 663-0148 663-1001 663-0149 663-0150 663-0271 663-0272 663-0273 663-0274 663-0231 663-0232 663-0266 663-0267 663-0310
Sensori di conducibilità
Elettrodi Conduttimetri
These conductivity probes with integrated temperature sensor (NTC 30 kΩ) are suitable for different sample types. They all provide automatic temperature compensation.
Suitable for:
InLab 710/InLab 720/InLab 725: SevenEasy™, SevenMulti™, FiveEasy™, FiveGo™
InLab 731/InLab 741: SevenExcellence™, SevenCompact™
InLab 737/InLab 738: SevenGo Duo™ SG23, SevenGo Duo pro™ SG78
InLab 742: SevenGo™ SG3, SevenGo pro™ SG7