Daptomycin
Fornitore: Thermo Fisher Scientific
461371000
461375000
ACRO461371000EA
144
EUR
ACRO461371000
ACRO461375000
Daptomycin
Daptomycin
Formula:
C₇₂H₁₀₁N₁₇O₂₆ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD08282794 CAS Number: 103060-53-3 |
Risultati del test di specificazione
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % |
Solubility | (0.5% in MeOH) Clear colorless to light yellow |
HPLC | ≥93 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...